missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-4-iodopyridine, 97%, Thermo Scientific Chemicals

Product Code. 11496880
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11496880 1 g 1g
11406890 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11496880 Supplier Thermo Scientific Alfa Aesar Supplier No. L20015.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 100523-96-4 | Molecular Formula: C5H3BrIN | Molecular Weight: 283.894 g/mol

It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Slightly soluble in water.

Notes
Light Sensitive. Store in dark. Store in cool, dry place in tightly closed container. Store away from oxidizing agent.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 100523-96-4
Molecular Formula C5H3BrIN
Molecular Weight (g/mol) 283.894
MDL Number MFCD03085768
InChI Key HPKRNLGLZYOVJS-UHFFFAOYSA-N
PubChem CID 1051520
IUPAC Name 2-bromo-4-iodopyridine
SMILES C1=CN=C(C=C1I)Br

Specifications

Melting Point 61°C
Quantity 1 g
Beilstein 108925
Sensitivity Light sensitive
Synonym 2-bromo-4-iodo-pyridine, pyridine, 2-bromo-4-iodo, 4-iodo-2-bromopyridine, pyridine,2-bromo-4-iodo, pubchem5956, acmc-20a0oj, ksc493g9n, abbypharma ap-13-5788
Solubility Information Slightly soluble in water.
Formula Weight 283.89
Percent Purity 97%
Chemical Name or Material 2-Bromo-4-iodopyridine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.