missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-4-fluorophenol, 98%, ACROS Organics™

Product Code. 10567753 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
:
1g; Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10567753 1g; Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10567753 Supplier Thermo Scientific Acros Supplier No. 311070010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 496-69-5
CAS Min % 97.5
CAS Max % 100.0
Melting Point 42°C to 46°C
Color Brown
Density 1.744g/mL
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Molecular Formula C6H4BrFO
Linear Formula BrC6H3(F)OH
MDL Number MFCD00010614
Quantity 1 g
Synonym 2-bromo-4-fluoro-phenol, phenol, 2-bromo-4-fluoro, 4-fluoro-2-bromo phenol, pubchem1485, 2-bromo-4-flourophenol, 2-bromo4-fluoro-phenol, acmc-1artu, 2-bromo-4-fluoro phenol, ksc490k9b, akos akm01586
InChI Key MEYRABVEYCFHHB-UHFFFAOYSA-N
SMILES C1=CC(=C(C=C1F)Br)O
IUPAC Name 2-bromo-4-fluorophenol
Molecular Weight (g/mol) 190.999
PubChem CID 605472
Formula Weight 191.01
Percent Purity ≥97.5%
Physical Form Crystalline Solid
Chemical Name or Material 2-Bromo-4-fluorophenol, 98%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.