missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-4-fluorophenol, 98+%

Product Code. 11463614
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11463614 100 g 100g
11443614 5 g 5g
11453614 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11463614 Supplier Thermo Scientific Alfa Aesar Supplier No. A11263.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 496-69-5 | C6H4BrFO | 190.999 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 496-69-5
Molecular Formula C6H4BrFO
Molecular Weight (g/mol) 190.999
MDL Number MFCD00010614
InChI Key MEYRABVEYCFHHB-UHFFFAOYSA-N
Synonym 2-bromo-4-fluoro-phenol, phenol, 2-bromo-4-fluoro, 4-fluoro-2-bromo phenol, pubchem1485, 2-bromo-4-flourophenol, 2-bromo4-fluoro-phenol, acmc-1artu, 2-bromo-4-fluoro phenol, ksc490k9b, akos akm01586
PubChem CID 605472
IUPAC Name 2-bromo-4-fluorophenol
SMILES C1=CC(=C(C=C1F)Br)O

Specifications

Melting Point 42°C to 46°C
Boiling Point 123°C to 125°C (50 mmHg)
Flash Point 85°C (185°F)
Quantity 100 g
Beilstein 2355593
Formula Weight 191
Percent Purity ≥98%
Chemical Name or Material 2-Bromo-4-fluorophenol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.