missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-4-fluorobenzoic acid, 98%

Product Code. 11374236
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11374236 5 g 5g
11364236 1 g 1g
11384236 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11374236 Supplier Thermo Scientific Alfa Aesar Supplier No. L19321.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1006-41-3 | C7H4BrFO2 | 219.009 g/mol

2-Bromo-4-fluorobenzoic acid is used in the synthesis of 3-fluoro-8-(methylthio)dibenzo[b,f]thiepin-10(11H)-one, 2-fluoro-8-(methylthio)dibenzo[b,f]thiepin-10(11H)-one, 2-((2-carboxy-5-fluorophenyl)amino)-3-methoxybenzoic acid and 2-bromo-4-fluorobenzamide. It is also used in the preparation of boron-containing anti-fungal agents, in the treatment of onychomycosis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Bromo-4-fluorobenzoic acid is used in the synthesis of 3-fluoro-8-(methylthio)dibenzo[b,f]thiepin-10(11H)-one, 2-fluoro-8-(methylthio)dibenzo[b,f]thiepin-10(11H)-one, 2-((2-carboxy-5-fluorophenyl)amino)-3-methoxybenzoic acid and 2-bromo-4-fluorobenzamide. It is also used in the preparation of boron-containing anti-fungal agents, in the treatment of onychomycosis.

Solubility
Soluble in water and methanol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1006-41-3
Molecular Formula C7H4BrFO2
Molecular Weight (g/mol) 219.009
MDL Number MFCD00055370
InChI Key RRKPMLZRLKTDQV-UHFFFAOYSA-N
Synonym 2-bromo-4-fluorobenzoicacid, 4-fluoro-2-bromobenzoic acid, 2-bromo-4-fluoro-benzoic acid, benzoic acid, 2-bromo-4-fluoro, rarechem al bo 1996, buttpark 18\02-85, 2-bromo-4-fluorbenzoic acid, pubchem1314, 2-bromo-4-fluorobenzoic, 2-bromofluorobenzoic acid
PubChem CID 1268169
IUPAC Name 2-bromo-4-fluorobenzoic acid
SMILES C1=CC(=C(C=C1F)Br)C(=O)O

Specifications

Melting Point 167°C to 169°C
Quantity 5 g
Beilstein 2614292
Solubility Information Soluble in water and methanol.
Formula Weight 219.01
Percent Purity 98%
Chemical Name or Material 2-Bromo-4-fluorobenzoic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.