missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-4-fluoroaniline, 98%

Product Code. 11409303
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11409303 100 g 100g
11489293 5 g 5g
11499293 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11409303 Supplier Thermo Scientific Alfa Aesar Supplier No. A11751.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 1003-98-1 | C6H5BrFN | 190.015 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1003-98-1
Molecular Formula C6H5BrFN
Molecular Weight (g/mol) 190.015
MDL Number MFCD00042462
InChI Key YLMFXCIATJJKQL-UHFFFAOYSA-N
Synonym 2-bromo-4-fluorobenzenamine, 4-fluoro-2-bromoaniline, 2-bromo-4-fluoro-aniline, benzenamine, 2-bromo-4-fluoro, 2-bromo-4-fluoro-phenylamine, 2-bromo-4-fluoro aniline, 2-bromo-4-fluorophenylamine, 2-brom-4-fluoranilin, bromofluoroaniline1, pubchem2918
PubChem CID 242873
IUPAC Name 2-bromo-4-fluoroaniline
SMILES C1=CC(=C(C=C1F)Br)N

Specifications

Density 1.67
Boiling Point 221°C
Flash Point 104°C (219°F)
Refractive Index 1.583
Quantity 100 g
UN Number UN2810
Beilstein 2802562
Formula Weight 190.01
Percent Purity 98%
Chemical Name or Material 2-Bromo-4-fluoroaniline

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.