missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-4-cyanopyridine, 97%

Product Code. 11416360
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11416360 5 g 5g
11406360 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11416360 Supplier Thermo Scientific Alfa Aesar Supplier No. H27191.06
 Restricted Item
Estimated Shipment: 07-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 10386-27-3 | C6H3BrN2 | 183.01 g/mol

Mainly bromo compounds find application as dyes. Also in pharmaceutical industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Mainly bromo compounds find application as dyes. Also in pharmaceutical industry.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10386-27-3
Molecular Formula C6H3BrN2
Molecular Weight (g/mol) 183.01
MDL Number MFCD07367879
InChI Key AWSJFEKOXQBDSL-UHFFFAOYSA-N
Synonym 2-bromo-4-cyanopyridine, 2-bromoisonicotinonitrile, 2-bromo-isoniconinonitrile, 4-pyridinecarbonitrile, 2-bromo, 2-bromo-isonicotinonitrile, 2-bromo-4-pyridinecarbonitrile, pubchem14366, 2-bromoisonicotnonitrile, 2-bromo-4-cyano pyridine, 2-bromo-4-cyano-pyridine
PubChem CID 11735544
IUPAC Name 2-bromopyridine-4-carbonitrile
SMILES BrC1=NC=CC(=C1)C#N

Specifications

Melting Point 70°C to 74°C
Odor Odorless
Quantity 5 g
Solubility Information Insoluble in water.
Formula Weight 183.01
Percent Purity 97%
Chemical Name or Material 2-Bromo-4-cyanopyridine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.