missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-4-chloroanisole, 98%, Thermo Scientific™

Product Code. 10191042 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
Packaging:
Glass bottle
Unit Size:
25mL
5mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10191042 5 mL Glass bottle 5mL
10762833 25 mL Glass bottle 25mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10191042 Supplier Thermo Scientific Acros Supplier No. 366560050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 60633-25-2
Molecular Formula C7H6BrClO
Molecular Weight (g/mol) 221.48
MDL Number MFCD00079705
InChI Key YJEMGEBDXDPBSP-UHFFFAOYSA-N
Synonym 2-bromo-4-chloroanisole, 2-bromo-4-chloro-1-methoxy-benzene, benzene, 2-bromo-4-chloro-1-methoxy, 4-chloro-2-bromoanisole, pubchem2629, 2-bromo4-chloroanisole, 4-chloro-2-bromanisole, 2-bromo-4-chloro anisole, acmc-1b9j6, ksc490k3r
PubChem CID 521935
IUPAC Name 2-bromo-4-chloro-1-methoxybenzene
SMILES COC1=C(C=C(C=C1)Cl)Br

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Colorless to Yellow
Density 1.6250g/mL
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula BrC6H3(Cl)OCH3
Refractive Index 1.5850 to 1.5870 (20°C, 589nm)
Quantity 5 mL
Specific Gravity 1.625
Formula Weight 221.48
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 2-Bromo-4-chloroanisole, 98%
Show More Show Less
compliance-icons
Product Identifier
  • 2-Bromo-4-chloroanisole
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.