missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-4-chloroaniline, 98%, Thermo Scientific™

Product Code. 10065153 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10065153 5 g Glass bottle 5g
10278803 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10065153 Supplier Thermo Scientific Acros Supplier No. 393470050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 873-38-1
Molecular Formula C6H5BrClN
Molecular Weight (g/mol) 206.47
MDL Number MFCD00041313
InChI Key SYTBIFURTZACKR-UHFFFAOYSA-N
Synonym benzenamine, 2-bromo-4-chloro, 1-amino-2-bromo-4-chlorobenzene, 2-bromo-4-chloro-phenylamine, 2-bromo-4-chloro aniline, 2-bromo-4-chlorophenylamine, pubchem3576, acmc-1bkdm, 2-bromo-4-chloro-aniline, 4-chloro-2-bromoaniline, 2-bromo-4-chloroaniline
PubChem CID 70110
IUPAC Name 2-bromo-4-chloroaniline
SMILES C1=CC(=C(C=C1Cl)Br)N

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 64°C to 68°C
Color Brown
Boiling Point 127°C (20.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula ClC6H3(Br)NH2
Quantity 5 g
Formula Weight 206.47
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 2-Bromo-4-chloroaniline
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 2-Bromo-4-chloroaniline
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H373-May cause damage to organs.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.