missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-4,6-dimethylaniline, 98%

Product Code. p-7025657
Click to view available options
Quantity:
10 g
25 g
50 g
Unit Size:
10g
25g
50g
This item is not returnable. View return policy

Product Code. 11449167

Brand: Thermo Scientific Alfa Aesar B21272.09

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 41825-73-4 | C8H10BrN | 200.08 g/mol

2-Bromo-4,6-dimethylaniline can react with 3-chloro-3-methyl-but-1-yne to produce 2-bromo-N-(1',1'-dimethylprop-2'-ynyl)-4,6-dimethylaniline. This reaction will need reagent Et3N, catalyst CuCl, and the menstruum dioxane.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Bromo-4,6-dimethylaniline can react with 3-chloro-3-methyl-but-1-yne to produce 2-bromo-N-(1′,1′-dimethylprop-2′-ynyl)-4,6-dimethylaniline. This reaction will need reagent Et3N, catalyst CuCl, and the menstruum dioxane.

Solubility
Insoluble in water.

Notes
Keep container tightly closed. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 41825-73-4
Molecular Formula C8H10BrN
Molecular Weight (g/mol) 200.08
MDL Number MFCD00047826
InChI Key YOSJCQJJIHEUKA-UHFFFAOYSA-N
Synonym 2-bromo-4,6-dimethyl-aniline, 6-bromo-2,4-xylidine, benzenamine, 2-bromo-4,6-dimethyl, 2-bromo-4,6-dimethylphenylamine, 4-amino-5-bromo-m-xylene, 2-bromo-4,6-dimethylbenzenamine, 2-bromo-4,6-dimethyl-phenylamine, 2-amino-3,5-dimethyl-1-bromobenzene, pubchem3761
PubChem CID 170579
IUPAC Name 2-bromo-4,6-dimethylaniline
SMILES CC1=CC(C)=C(N)C(Br)=C1

Specifications

Melting Point 42°C to 46°C
Boiling Point 75°C to 79°C (5 mmHg)
Quantity 10 g
UN Number UN2811
Beilstein 3046484
Solubility Information Insoluble in water.
Formula Weight 200.08
Percent Purity 98%
Chemical Name or Material 2-Bromo-4,6-dimethylaniline

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.