missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-4,6-difluoroaniline, 98%

Product Code. 11438853
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
This item is not returnable. View return policy

Product Code. 11438853

Brand: Thermo Scientific Alfa Aesar A11022.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 444-14-4 | C6H4BrF2N | 208.01 g/mol

2-Bromo-4,6-difluoroaniline is used in chemical synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Bromo-4,6-difluoroaniline is used in chemical synthesis.

Solubility
Insoluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 444-14-4
Molecular Formula C6H4BrF2N
Molecular Weight (g/mol) 208.01
MDL Number MFCD00009639
InChI Key WUJKFVGKLTWVSQ-UHFFFAOYSA-N
Synonym 2,4-difluoro-6-bromoaniline, 6-bromo-2,4-difluoroaniline, 2-bromo-4,6-difluoro-phenylamine, 2-bromo-4,6-difluorobenzenamine, benzenamine, 2-bromo-4,6-difluoro, 2-bromo-4,6-difluorophenylamine, pubchem2913, acmc-1ao2h, 2-bromo4,6-difluoroaniline
PubChem CID 136285
IUPAC Name 2-bromo-4,6-difluoroaniline
SMILES NC1=C(F)C=C(F)C=C1Br

Specifications

Melting Point 38°C to 42°C
Odor Weak, characteristic
Quantity 25 g
UN Number UN3077
Beilstein 2718139
Solubility Information Insoluble in water.
Formula Weight 208.01
Percent Purity 98%
Chemical Name or Material 2-Bromo-4,6-difluoroaniline

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.