missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-4,6-di-tert-butylphenol, 97%

Product Code. 11440733
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11440733 25 g 25g
11450733 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11440733 Supplier Thermo Scientific Alfa Aesar Supplier No. A13218.14
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 20834-61-1 | C14H21BrO | 285.225 g/mol

It is used in pharmaceutical intermediates and as medicine. When 2-Bromo-4,6-di-tert-butylphenol was treated with potassium ferricyanide in benzene 1,4-dihydro-4-bromo-2,4,6,8-tetra-tert-butyl-1-oxodibenzofuran was yielded with a small amount of 2,4,6,8-tetra-tert-butyldibenzofuran.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in pharmaceutical intermediates and as medicine. When 2-Bromo-4,6-di-tert-butylphenol was treated with potassium ferricyanide in benzene 1,4-dihydro-4-bromo-2,4,6,8-tetra-tert-butyl-1-oxodibenzofuran was yielded with a small amount of 2,4,6,8-tetra-tert-butyldibenzofuran.

Notes
Store at room temperature. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 20834-61-1
Molecular Formula C14H21BrO
Molecular Weight (g/mol) 285.225
MDL Number MFCD00051597
InChI Key DIWZVAHZEOFSLS-UHFFFAOYSA-N
Synonym 2-bromo-4,6-di-tert-butylphenol, 2-bromo-4,6-di-t-butylphenol, 2,4-di-tert-butyl-6-bromophenol, phenol, 2-bromo-4,6-bis 1,1-dimethylethyl, phenol,2-bromo-4,6-bis 1,1-dimethylethyl, acmc-1crlp, 4,6-di-t-butyl-2-bromophenol, 6-bromo-2,4-di-t-butylphenol, 2-bromo4,6-di-tert-butylphenol, 2-bromo-4, 6-di-tert-butylphenol
PubChem CID 519822
IUPAC Name 2-bromo-4,6-ditert-butylphenol
SMILES CC(C)(C)C1=CC(=C(C(=C1)Br)O)C(C)(C)C

Specifications

Melting Point 55°C to 59°C
Quantity 25 g
Beilstein 2100150
Formula Weight 285.23
Percent Purity 97%
Chemical Name or Material 2-Bromo-4,6-di-tert-butylphenol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.