missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-4,5-dimethoxybenzoic acid, 97%, Thermo Scientific™

Product Code. 10193064 Shop All Thermo Scientific Chemicals Products
Encompass
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10193064 1 g Glass bottle 1g
10294594 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10193064 Supplier Thermo Scientific Acros Supplier No. 395090010

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 6286-46-0
Molecular Formula C9H9BrO4
Molecular Weight (g/mol) 261.07
MDL Number MFCD00017542
InChI Key HWFCHCRFQWEFMU-UHFFFAOYSA-N
Synonym 6-bromoveratric acid, benzoic acid, 2-bromo-4,5-dimethoxy, 2-bromo-4,5-dimethoxybenzoicacid, 6-bromo-3,4-dimethoxybenzoic acid, 6-bromveratrumsaure, acmc-1b7v8, ksc496e4l, rarechem al be 0015, timtec-bb sbb037916
PubChem CID 222963
IUPAC Name 2-bromo-4,5-dimethoxybenzoic acid
SMILES COC1=CC(Br)=C(C=C1OC)C(O)=O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 188°C to 190°C
Color Cream to White or Yellow
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Linear Formula (CH3O)2C6H2(Br)CO2H
Quantity 1 g
Formula Weight 261.07
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 2-Bromo-4, 5-dimethoxybenzoic acid
Show More Show Less
compliance-icons
Product Identifier
  • 2-Bromo-4,5-dimethoxybenzoic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.