missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-4,5-difluorobenzoic acid, 97%

Product Code. 10507254 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10507254 25 g Glass bottle 25g
10376394 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10507254 Supplier Thermo Scientific Acros Supplier No. 399040250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 64695-84-7 | C7H3BrF2O2 | 237 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 64695-84-7
Molecular Formula C7H3BrF2O2
Molecular Weight (g/mol) 237
MDL Number MFCD04973422
InChI Key DGCBGBZYTNTZJH-UHFFFAOYSA-N
Synonym 2-bromo-4,5-difluorobenzoicacid, 2-bromo-4,5-difluoro-benzoic acid, benzoic acid, 2-bromo-4,5-difluoro, pubchem4991, acmc-209nnq, ksc495k3l, benzoic acid,2-bromo-4,5-difluoro, 2-bromo-4,5-difluorobenzoic acid, 2-bromanyl-4,5-bis fluoranyl benzoic acid
PubChem CID 2782789
IUPAC Name 2-bromo-4,5-difluorobenzoic acid
SMILES C1=C(C(=CC(=C1F)F)Br)C(=O)O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 118.0°C to 122.0°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Linear Formula F2C6H2(Br)CO2H
Quantity 25 g
Formula Weight 237
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 2-Bromo-4, 5-difluorobenzoic acid
Show More Show Less
compliance-icons
Product Identifier
  • 2-Bromo-4,5-difluorobenzoic acid
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.