missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-Bromo-4,5-difluorobenzoic acid, 97%

Product Code. 15436888
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15436888 25 g 25g
15426888 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15436888 Supplier Alfa Aesar™ Supplier No. H60418.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 64695-84-7
Molecular Formula C7H3BrF2O2
Molecular Weight (g/mol) 237
MDL Number MFCD04973422
InChI Key DGCBGBZYTNTZJH-UHFFFAOYSA-N
PubChem CID 2782789
IUPAC Name 2-bromo-4,5-difluorobenzoic acid
SMILES C1=C(C(=CC(=C1F)F)Br)C(=O)O

Specifications

Melting Point 118°C to 122°C
Quantity 25 g
Synonym 2-bromo-4,5-difluorobenzoicacid, 2-bromo-4,5-difluoro-benzoic acid, benzoic acid, 2-bromo-4,5-difluoro, pubchem4991, acmc-209nnq, ksc495k3l, benzoic acid,2-bromo-4,5-difluoro, 2-bromo-4,5-difluorobenzoic acid, 2-bromanyl-4,5-bis fluoranyl benzoic acid
Formula Weight 237
Percent Purity 97%
Chemical Name or Material 2-Bromo-4,5-difluorobenzoic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.