missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-4,5-dichloro-1H-imidazole, 97%, Thermo Scientific™

Product Code. 10354472
Change view
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10354472 10 g Amber Glass Bottle 10g
10466462 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10354472 Supplier Thermo Scientific Maybridge Supplier No. KM01467EA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 16076-27-0
Molecular Formula C3HBrCl2N2
Molecular Weight (g/mol) 215.86
MDL Number MFCD00067894
InChI Key SULQOTGFPPFUMG-UHFFFAOYSA-N
Synonym 2-bromo-4,5-dichloroimidazole, imidazole, 2-bromo-4,5-dichloro, 1h-imidazole, 2-bromo-4,5-dichloro, imidazole,5-dichloro, acmc-20ao2s, 1h-imidazole,5-dichloro, 5-23-04-00463 beilstein handbook reference, 2-bromo-4,5-dichloro-imidazole, 2-bromo-4,5-dichloro-3h-imidazole
PubChem CID 99484
IUPAC Name 2-bromo-4,5-dichloro-1H-imidazole
SMILES ClC1=C(Cl)N=C(Br)N1

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 10 g
Formula Weight 215.86
Percent Purity 97%
Chemical Name or Material 2-Bromo-4, 5-dichloro-1H-imidazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.