missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-4,4,4-trifluorobutyric acid ethyl ester, Thermo Scientific™

Product Code. 10057889
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10057889 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10057889 Supplier Thermo Scientific Maybridge Supplier No. MAY00179DA
 Restricted Item
Estimated Shipment: 06-11-2026
Add to basket
This item is not returnable. View return policy

Specifications

CAS 367-33-9
Assay Percent Range 97+%
Packaging Amber Glass Bottle
Molecular Formula C6H8BrF3O2
Quantity 1 g
Synonym 2-bromo-4,4,4-trifluorobutyric acid ethyl ester, ethyl 2-bromo-4,4,4-trifluorobutyrate, ethyl 2-bromo-4,4,4-trifluoro-butanoate, butanoic acid,2-bromo-4,4,4-trifluoro-, ethyl ester
InChI Key POVYUFTVLGOXEE-UHFFFAOYSA-N
SMILES CCOC(=O)C(CC(F)(F)F)Br
IUPAC Name ethyl 2-bromo-4,4,4-trifluorobutanoate
Molecular Weight (g/mol) 249.027
PubChem CID 45594306
Formula Weight 249.03
Percent Purity 97%
Chemical Name or Material 2-Bromo-4, 4, 4-trifluorobutyric acid ethyl ester
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.