missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-4'-(1-pyrrolidinyl)acetophenone, 97%

Product Code. 11318266
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11318266 25 g 25g
11308266 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11318266 Supplier Thermo Scientific Alfa Aesar Supplier No. L15407.14
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 216144-18-2 | C12H14BrNO | 268.154 g/mol

2-Bromo-4'-(1-pyrrolidinyl)acetophenone is an important raw material and used as pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Bromo-4′-(1-pyrrolidinyl)acetophenone is an important raw material and used as pharmaceutical intermediates.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 216144-18-2
Molecular Formula C12H14BrNO
Molecular Weight (g/mol) 268.154
MDL Number MFCD01075690
InChI Key WLRIUCQUMKJOGT-UHFFFAOYSA-N
Synonym 2-bromo-1-4-pyrrolidin-1-yl phenyl ethanone, alpha-bromo-4-1-pyrrolidino acetophenone, 2-bromo-1-4-pyrrolidin-1-ylphenyl ethanone, 2-bromo-1-4-1-pyrrolidinyl phenyl ethanone, ethanone, 2-bromo-1-4-1-pyrrolidinyl phenyl, 2-bromo-1-4-pyrrolidin-1-yl phenyl ethan-1-one, 2-bromo-1-4-pyrrolidin-1-ylphenyl ethan-1-one, 4-pyrrolidin-1-yl phenacyl bromide, 2-bromo-1-4-pyrrolidinylphenyl ethan-1-one, acmc-209fm8
PubChem CID 3681785
IUPAC Name 2-bromo-1-(4-pyrrolidin-1-ylphenyl)ethanone
SMILES C1CCN(C1)C2=CC=C(C=C2)C(=O)CBr

Specifications

Melting Point 120°C to 122°C
Quantity 25 g
UN Number UN3261
Beilstein 8549816
Solubility Information Insoluble in water.
Formula Weight 268.16
Percent Purity 97%
Chemical Name or Material 2-Bromo-4'-(1-pyrrolidinyl)acetophenone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.