missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-3'-(trifluoromethyl)acetophenone, 98%

Product Code. 15434127
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15434127 5 g 5g
11325145 1 g 1g
11315145 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15434127 Supplier Thermo Scientific Alfa Aesar Supplier No. H31526.06
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 2003-10-3 | C9H6BrF3O | 267.05 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2003-10-3
Molecular Formula C9H6BrF3O
Molecular Weight (g/mol) 267.05
MDL Number MFCD03094283
InChI Key TZIYNLSEBAYCBZ-UHFFFAOYSA-N

Specifications

Melting Point 20°C to 23°C
Boiling Point 69°C to 72°C
Quantity 5 g
Synonym 2-bromo-1-3-trifluoromethyl phenyl ethanone, 3-trifluoromethyl phenacyl bromide, 2-bromo-3'-trifluoromethylacetophenone, 2-bromo-1-3-trifluoromethyl phenyl ethan-1-one, 2-bromo-3'-trifluoromethyl acetophenone, 2-bromo-1-3-trifluoromethyl-phenyl-ethanone, 2-bromo-1-3-trifluoromethyl phenyl-1-ethanone, ethanone, 2-bromo-1-3-trifluoromethyl phenyl, 3-trifluoromethylphenacylbromide
SMILES C1=CC(=CC(=C1)C(F)(F)F)C(=O)CBr
IUPAC Name 2-bromo-1-[3-(trifluoromethyl)phenyl]ethanone
PubChem CID 2778386
Formula Weight 267.04
Percent Purity 98%
Chemical Name or Material 2-Bromo-3'-(trifluoromethyl)acetophenone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.