missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-3-pyridinecarboxaldehyde, 96%

Product Code. 10294664 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10294664 1 g Glass bottle 1g
10515334 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10294664 Supplier Thermo Scientific Acros Supplier No. 399670010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 128071-75-0 | C6H4BrNO | 186 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 128071-75-0
Molecular Formula C6H4BrNO
Molecular Weight (g/mol) 186
MDL Number MFCD04966945
InChI Key GNFWMEFWZWXLIN-UHFFFAOYSA-N
Synonym 2-bromonicotinaldehyde, 2-bromo-3-formylpyridine, 2-bromo-3-pyridinecarboxaldehyde, 2-bromopyridine-3-carboxaldehyde, 3-pyridinecarboxaldehyde, 2-bromo, pubchem5057, acmc-1bypv, 2-bromo-3-formyl pyridine, bromo-3-pyridine carbaldehyde, bromo-3-pyridinecarboxaldehyde
PubChem CID 4714951
IUPAC Name 2-bromopyridine-3-carbaldehyde
SMILES C1=CC(=C(N=C1)Br)C=O

Specifications

CAS Min % 95.0
CAS Max % 100.0
Melting Point 70.0°C to 74.0°C
Boiling Point 100.0°C (3.0 mmHg)
Assay Percent Range 95% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 186
Percent Purity 96%
Chemical Name or Material 2-Bromo-3-pyridinecarboxaldehyde
compliance-icons
Product Identifier
  • 2-Bromo-3-pyridinecarboxaldehyde
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.