missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-3'-nitroacetophenone, 97%

Product Code. 10501564 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10501564 5 g Glass bottle 5g
10655572 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10501564 Supplier Thermo Scientific Acros Supplier No. 137470050
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 2227-64-7 | C8H6BrNO3 | 244.04 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2227-64-7
Molecular Formula C8H6BrNO3
Molecular Weight (g/mol) 244.04
MDL Number MFCD00024512
InChI Key GZHPNIQBPGUSSX-UHFFFAOYSA-N
Synonym 2-bromo-1-3-nitrophenyl ethanone, 2-bromo-3'-nitroacetophenone, 3-nitrophenacyl bromide, 3-nitrophenacylbromide, 2-bromo-1-3-nitrophenyl ethan-1-one, a-bromo-3-nitroacetophenone, 2-bromo-3-nitroacetophenone, alpha-bromo-3'-nitroacetophenone, acetophenone, 2-bromo-3'-nitro, m-nitrophenacyl bromide
PubChem CID 75213
IUPAC Name 2-bromo-1-(3-nitrophenyl)ethanone
SMILES C1=CC(=CC(=C1)[N+](=O)[O-])C(=O)CBr

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 90°C to 94°C
Color Beige-Orange to Green-Yellow
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula O2NC6H4COCH2Br
Quantity 5 g
Solubility Information Solubility in water: insoluble
Formula Weight 244.04
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 2-Bromo-3'-nitroacetophenone, 97%
Show More Show Less
compliance-icons
Product Identifier
  • 2-Bromo-3'-nitroacetophenone
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.