missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-3-methylbutyrylurea, TRC

Product Code. 30136623
Change view
Click to view available options
Quantity:
1 g
10 g
50 g
Unit Size:
10g
1g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30136623 50 g 50g
30132741 10 g 10g
30005869 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30136623 Supplier Toronto Research Chemicals Supplier No. TRCB68513050G
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 496-67-3
Molecular Formula C6 H11 Br N2 O2
Molecular Weight (g/mol) 223.07
Synonym Butanamide, N-(aminocarbonyl)-2-bromo-3-methyl-, Urea, (2-bromo-3-methylbutyryl)- (6CI,8CI), (RS)-2-Bromoisovalerylurea, (+/-)-Bromisoval, (alpha-Bromoisovaleryl)urea, 2-Bromo-3-methylbutyrylurea, 2-Bromoisovalerylurea, Abroval, Alluval, Alural, BVU, Bromaral, Bromcarbamide, Bromisoval, Bromisovalerylurea, Bromisovalum, Bromizoval, Bromocarbamide, Bromoisovalum, Bromoval, Bromovalerocarbamide, Bromovaleroylurea, Bromovalerylurea, Bromoxil, Bromural, Bromuvan, Bromvalerylurea, Bromvaletone, Bromvalurea, Bromyl, Brovalin, Brovalurea, Brovarin, Calmotin, Dagrabromyl, Dibroluur, Dormigene, Isobromyl, Pivadorm, Pivadorn, Somnurol, Upiol, Uvaleral, alpha-Bromisovalerylurea, alpha-Bromo-beta-dimethylpropanoylurea, alpha-Bromoisovaleric acid ureide, alpha-Bromoisovaleroylurea
IUPAC Name 2-bromo-N-carbamoyl-3-methylbutanamide
SMILES CC(C)C(Br)C(=O)NC(=O)N

Specifications

Quantity 50 g
InChI Formula InChI=1S/C6H11BrN2O2/c1-3(2)4(7)5(10)9-6(8)11/h3-4H,1-2H3,(H3,8,9,10,11)
Formula Weight 222.0004
Chemical Name or Material Bromisoval
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.