missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-3-methylbutyric acid, 98%, Thermo Scientific™

Product Code. 10666471 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10666471 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10666471 Supplier Thermo Scientific Acros Supplier No. 268880250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 565-74-2
CAS Min % 97.5
CAS Max % 100.0
Melting Point 39.0°C to 43.0°C
Color White to Beige
Boiling Point 124.0°C to 126.0°C (20.0 mmHg)
Flash Point 107°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Molecular Formula C5H9BrO2
Linear Formula (CH3)2CHCH(Br)CO2H
MDL Number MFCD00004210
Quantity 25 g
Beilstein 02, 317
Merck Index 15, 1432
Synonym 2-bromo-3-methylbutyric acid, 2-bromoisovaleric acid, butanoic acid, 2-bromo-3-methyl, alpha-bromoisovaleric acid, butyric acid, 2-bromo-3-methyl, 2-bromo-3-methyl-butanoic acid, .alpha.-bromoisovaleric acid, +/--2-bromo-3-methylbutyric acid, bromisovaleriansaure, acmc-209wpb
InChI Key UEBARDWJXBGYEJ-UHFFFAOYSA-N
SMILES CC(C)C(C(=O)O)Br
IUPAC Name 2-bromo-3-methylbutanoic acid
Molecular Weight (g/mol) 181.03
PubChem CID 11268
Formula Weight 181.03
Percent Purity 98%
Physical Form Crystalline Powder and/or Chunks
Chemical Name or Material 2-Bromo-3-methylbutyric acid, 98%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2-Bromo-3-methylbutyric acid
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H312-Harmful in contact with skin.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.