missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-3'-methoxyacetophenone, 97%

Product Code. 10695582 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
25 g
Packaging:
Glass bottle
Unit Size:
10g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10695582 10 g Glass bottle 10g
10605592 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10695582 Supplier Thermo Scientific Acros Supplier No. 151910100

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 5000-65-7 | C9H9BrO2 | 229.07 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5000-65-7
Molecular Formula C9H9BrO2
Molecular Weight (g/mol) 229.07
MDL Number MFCD00000199
InChI Key IOOHBIFQNQQUFI-UHFFFAOYSA-N
Synonym 2-bromo-1-3-methoxyphenyl ethanone, 2-bromo-3'-methoxyacetophenone, m-methoxyphenacyl bromide, 3'-methoxyphenacyl bromide, 3-bromoacetyl anisole, 2-bromo-1-3-methoxyphenyl ethan-1-one, 3-methoxyphenacyl bromide, bromomethyl 3-methoxyphenyl ketone, ccris 7998, 3-methoxyphenacylbromide
PubChem CID 101294
IUPAC Name 2-bromo-1-(3-methoxyphenyl)ethanone
SMILES COC1=CC=CC(=C1)C(=O)CBr

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 59°C to 64°C
Color Green-Yellow to White
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Linear Formula CH3OC6H4COCH2Br
Quantity 10 g
Formula Weight 229.07
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 2-Bromo-3'-methoxyacetophenone, 97%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2-Bromo-3'-methoxyacetophenone
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.