missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-3-hexylthiophene, 98%, Thermo Scientific Chemicals

Product Code. 15412698
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15412698 5 g 5g
15422698 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15412698 Supplier Thermo Scientific Alfa Aesar Supplier No. H56243.06
 Restricted Item
Estimated Shipment: 14-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 69249-61-2 | Molecular Formula: C10H15BrS | Molecular Weight: 247.194 g/mol

2-Bromo-3-hexylthiophene is used in the synthesis of End-capped Regioregular Poly(3-hexylthiophene). It is also used to synthesize well-defined head-to-tail-type oligothiophenes which are used in a number of high-technology applications including OLEDs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Bromo-3-hexylthiophene is used in the synthesis of End-capped Regioregular Poly(3-hexylthiophene). It is also used to synthesize well-defined head-to-tail-type oligothiophenes which are used in a number of high-technology applications including OLEDs.

Solubility
Slightly miscible with water.

Notes
Incompatible with strong oxidizing agentsstrong acids and bases.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 69249-61-2
Molecular Formula C10H15BrS
Molecular Weight (g/mol) 247.194
MDL Number MFCD09907959
InChI Key XQJNXCHDODCAJF-UHFFFAOYSA-N
Synonym 2-bromo-3-hexyl-thiophene, thiophene, 2-bromo-3-hexyl, acmc-209o6r, 2-bromo-3-hexyl thiophene, 2-bromo-3-n-hexylthiophene, amth047, 2-bromanyl-3-hexyl-thiophene, ksc491i5d, 2-bromo-3-hexylthiophene
PubChem CID 10960141
IUPAC Name 2-bromo-3-hexylthiophene
SMILES CCCCCCC1=C(SC=C1)Br

Specifications

Density 1.24
Flash Point 110°C (230°F)
Refractive Index 1.529
Quantity 5 g
Solubility Information Slightly miscible with water.
Formula Weight 247.2
Percent Purity 98%
Chemical Name or Material 2-Bromo-3-hexylthiophene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.