missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-3-furoic acid, 97%, Thermo Scientific™

Product Code. 10612305
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10612305 1 g Amber Glass Bottle 1g
10265414 5 g Amber Glass Bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10612305 Supplier Thermo Scientific Maybridge Supplier No. CC64701DA

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 197846-05-2
Molecular Formula C5H3BrO3
Molecular Weight (g/mol) 190.98
MDL Number MFCD09702417
InChI Key UJNWWTUUYCCIFL-UHFFFAOYSA-N
PubChem CID 21162497
IUPAC Name 2-bromofuran-3-carboxylic acid
SMILES C1=COC(=C1C(=O)O)Br

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Synonym 2-bromo-3-furoic acid, 2-bromo-3-carboxyfuran, 2-bromo3-furancarboxylic acid, 2-bromo-3-furancarboxylic acid, 3-furancarboxylic acid,2-bromo, 2-bromanylfuran-3-carboxylic acid, 3-furancarboxylic acid, 2-bromo, 2-bromofuran-3-carboxylic acid
Formula Weight 190.98
Percent Purity 97%
Chemical Name or Material 2-Bromo-3-furoic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.