missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-3-fluorotoluene, 98%, Thermo Scientific Chemicals

Product Code. 15438358
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15438358 25 g 25g
15448358 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15438358 Supplier Thermo Scientific Alfa Aesar Supplier No. H61785.14
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 59907-13-0 | Molecular Formula: C7H6BrF | Molecular Weight: 189.027 g/mol

2-Bromo-3-fluorotoluene is a important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Bromo-3-fluorotoluene is a important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.

Solubility
Slightly soluble in water.

Notes
Store in cool, well ventilated area. Keep container tightly closed. Store away from oxidizing agent.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 59907-13-0
Molecular Formula C7H6BrF
Molecular Weight (g/mol) 189.027
MDL Number MFCD08458010
InChI Key FYCXRRYRNRDSRM-UHFFFAOYSA-N
Synonym 2-bromo-3-fluorotoluene, 3-fluoro-2-bromo toluene, benzene, 2-bromo-1-fluoro-3-methyl, 2-brom-1-fluor-3-methylbenzol, pubchem4518, acmc-1awko, ksc493o4f, 2-bromo-1-fluoro-3-methyl-benzene
PubChem CID 108856
IUPAC Name 2-bromo-1-fluoro-3-methylbenzene
SMILES CC1=C(C(=CC=C1)F)Br

Specifications

Melting Point 118°C to 123°C
Density 1.503
Flash Point 76°C (169°F)
Refractive Index 1.533
Quantity 25 g
Solubility Information Slightly soluble in water.
Formula Weight 189.02
Percent Purity 98%
Chemical Name or Material 2-Bromo-3-fluorotoluene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.