missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-3-fluorobenzyl bromide, 98%

Product Code. 15450429
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Product Code. Quantity unitSize
15450429 5 g 5g
15440429 1 g 1g
2 options
This item is not returnable. View return policy

Product Code. 15450429

Brand: Thermo Scientific Alfa Aesar H63632.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1184918-22-6 | C7H5Br2F | 267.92 g/mol

It is employed as a intermediate for pharmaceutical.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as a intermediate for pharmaceutical.

Solubility
Slightly soluble in water.

Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1184918-22-6
Molecular Formula C7H5Br2F
Molecular Weight (g/mol) 267.92
MDL Number MFCD12911576
InChI Key ZJSRIMJDFLFPJI-UHFFFAOYSA-N
Synonym 2-bromo-1-bromomethyl-3-fluorobenzene, 2-bromo-3-fluorobenzyl bromide, 2-bromo-3-bromomethyl-1-fluorobenzene, 2-bromo-3-fluorobenzylbromide
PubChem CID 44181799
IUPAC Name 2-bromo-1-(bromomethyl)-3-fluorobenzene
SMILES FC1=CC=CC(CBr)=C1Br

Specifications

Quantity 5 g
UN Number UN3261
Solubility Information Slightly soluble in water.
Formula Weight 267.92
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 2-Bromo-3-fluorobenzyl bromide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.