missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-3-fluorobenzaldehyde, 98%, Thermo Scientific Chemicals

Product Code. 15481589
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 891180-59-9
Molecular Formula C7H4BrFO
Molecular Weight (g/mol) 203.01
MDL Number MFCD07782044
InChI Key CYJBMGYWRHGZBR-UHFFFAOYSA-N
Synonym 2-bromo-3-fluoro-benzaldehyde, benzaldehyde, 2-bromo-3-fluoro, pubchem1417, pubchem22500, acmc-1bwut, ksc494e8j, 6-fluoro-2-formylbromobenzene
PubChem CID 24820504
IUPAC Name 2-bromo-3-fluorobenzaldehyde
SMILES C1=CC(=C(C(=C1)F)Br)C=O

Specifications

Quantity 5 g
Beilstein 10529128
Sensitivity Light sensitive
Formula Weight 203.01
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 2-Bromo-3-fluorobenzaldehyde

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'