missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-3-fluoroaniline, 97%

Product Code. p-7053919
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
This item is not returnable. View return policy

Product Code. 15408268

Brand: Thermo Scientific Alfa Aesar H61687.MD

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 111721-75-6 | C6H5BrFN | 190.015 g/mol

2-Bromo-3-fluoroaniline is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Bromo-3-fluoroaniline is used as pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 111721-75-6
Molecular Formula C6H5BrFN
Molecular Weight (g/mol) 190.015
MDL Number MFCD07369915
InChI Key XZRSXRUYZXBTGD-UHFFFAOYSA-N
Synonym 2-bromo-3-fluorophenylamine, 2-bromo-3-fluoro-phenylamine, 2-bromo-3-fluorobenzenamine, benzenamine, 2-bromo-3-fluoro, pubchem1523, acmc-1c6kb, 2-bromo-3-fluoro-aniline, ksc493i1t, 2-bromo-3-fluoro-phenyl-amine
PubChem CID 14770005
IUPAC Name 2-bromo-3-fluoroaniline
SMILES C1=CC(=C(C(=C1)F)Br)N

Specifications

Color White
Density 1.670 g/mL
Flash Point 94°C (201°F)
Refractive Index 1.583
Quantity 250 mg
UN Number UN2811
Solubility Information Slightly soluble in water.
Formula Weight 190.01
Percent Purity 97%
Physical Form Crystalline
Chemical Name or Material 2-Bromo-3-fluoroaniline
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.