missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-3'-chloropropiophenone, 98%

Product Code. 15492439
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15492439 5 g 5g
15482439 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15492439 Supplier Thermo Scientific Alfa Aesar Supplier No. H64999.06
 Restricted Item
Estimated Shipment: 24-09-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 34911-51-8 | C9H8BrClO | 247.52 g/mol

It used as an important intermediate for raw material and intermediate used in organic synthesis agrochemical, pharmaceutical and dyestuff field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It used as an important intermediate for raw material and intermediate used in organic synthesis agrochemical, pharmaceutical and dyestuff field.

Solubility
Soluble in acetonitrile, chloroform, dichloromethane and ethyl acetate.

Notes
Refrigerate. Keep container tightly closed. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 34911-51-8
Molecular Formula C9H8BrClO
Molecular Weight (g/mol) 247.52
MDL Number MFCD03701150
InChI Key OFNMQTRHMBQQEA-UHFFFAOYNA-N
PubChem CID 4992116
IUPAC Name 2-bromo-1-(3-chlorophenyl)propan-1-one
SMILES CC(Br)C(=O)C1=CC(Cl)=CC=C1

Specifications

Color Colorless to Yellow
Density 1.532 g/mL
Refractive Index 1.577
Quantity 5 g
Beilstein 2360404
Synonym 2-bromo-3'-chloropropiophenone, 2-bromo-1-3-chlorophenyl propan-1-one, 2-bromo-1-3-chlorophenyl-1-propanone, 3'-chloro-2-bromopropiophenone, 1-propanone,2-bromo-1-3-chlorophenyl, 1-propanone, 2-bromo-1-3-chlorophenyl, 2-bromo-1-3-chloro-phenyl-propan-1-one, bromochlorophenylpropanone, bromo-3'-chloropropiophenone, ksc573e4l
Solubility Information Soluble in acetonitrile,chloroform,dichloromethane and ethyl acetate.
Formula Weight 247.52
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 2-Bromo-3'-chloropropiophenone
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.