missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-3-(chloromethyl)pyridine, 96%, Thermo Scientific™

Product Code. 15488947
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 15488947

Brand: Thermo Scientific Alfa Aesar H52313.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 944906-87-0
Molecular Formula C6H5BrClN
Molecular Weight (g/mol) 206.467
MDL Number MFCD10697618
InChI Key OWRYEOOYOHXZJX-UHFFFAOYSA-N
Synonym 2-bromo-3-chloromethyl pyridine, pyridine, 2-bromo-3-chloromethyl, 2-bromo-3-chloromethylene pyridine
PubChem CID 53416632
IUPAC Name 2-bromo-3-(chloromethyl)pyridine
SMILES C1=CC(=C(N=C1)Br)CCl

Specifications

Melting Point 34°C to 36°C
Quantity 1 g
Beilstein 15651009
Formula Weight 206.47
Percent Purity 96%
Chemical Name or Material 2-Bromo-3-(chloromethyl)pyridine
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.