missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-3-chloro-5-(trifluoromethyl)pyridine, 97%, Thermo Scientific™

Product Code. p-2697943
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
This item is not returnable. View return policy

Product Code. 10130672

Brand: Thermo Scientific Maybridge BTB09453DA

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 75806-84-7
Molecular Formula C6H2BrClF3N
Molecular Weight (g/mol) 260.438
MDL Number MFCD00153072
InChI Key SMTKGMYGLYWNDL-UHFFFAOYSA-N
Synonym 2-bromo-3-chloro-5-trifluoromethyl pyridine, librarion l938, abbypharma ap-13-5059, pyridine, 2-bromo-3-chloro-5-trifluoromethyl, pubchem3000, acmc-1bicf, ksc498c5d, buttpark 153\33-66, 2-bromo-3-chloro-5 trifluoromethyl pyridine
PubChem CID 2736237
IUPAC Name 2-bromo-3-chloro-5-(trifluoromethyl)pyridine
SMILES C1=C(C=NC(=C1Cl)Br)C(F)(F)F

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 1 g
Formula Weight 260.44
Percent Purity 97%
Chemical Name or Material 2-Bromo-3-chloro-5-(trifluoromethyl)pyridine
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.