missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-3-chloro-5-(trifluoromethyl)pyridine, 97%, Thermo Scientific™

Product Code. 10448894
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10448894 5 g 5g
10130672 1 g 1g
10011134 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10448894 Supplier Thermo Scientific Maybridge Supplier No. BTB09453DE
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 75806-84-7
Molecular Formula C6H2BrClF3N
Molecular Weight (g/mol) 260.438
MDL Number MFCD00153072
InChI Key SMTKGMYGLYWNDL-UHFFFAOYSA-N
Synonym 2-bromo-3-chloro-5-trifluoromethyl pyridine, librarion l938, abbypharma ap-13-5059, pyridine, 2-bromo-3-chloro-5-trifluoromethyl, pubchem3000, acmc-1bicf, ksc498c5d, buttpark 153\33-66, 2-bromo-3-chloro-5 trifluoromethyl pyridine
PubChem CID 2736237
IUPAC Name 2-bromo-3-chloro-5-(trifluoromethyl)pyridine
SMILES C1=C(C=NC(=C1Cl)Br)C(F)(F)F

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Quantity 5 g
Formula Weight 260.44
Percent Purity 97%
Chemical Name or Material 2-Bromo-3-chloro-5-(trifluoromethyl)pyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.