missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-3-(bromomethyl)pyridine, 96%, Thermo Scientific Chemicals

Catalog Number 15478947
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
94446-97-6
C6H5Br2N
250.921
MFCD08276299
NEWLOSCWMVKPGI-UHFFFAOYSA-N
2-bromo-3-bromomethyl pyridine, 2-bromo-3-bromomethylene pyridine, pubchem22284, 2-bromo-3-bromomethyl-pyridine, pyridine, 2-bromo-3-bromomethyl
10901078
2-bromo-3-(bromomethyl)pyridine
C1=CC(=C(N=C1)Br)CBr
This item is not returnable. View return policy
94446-97-6
C6H5Br2N
250.921
MFCD08276299
NEWLOSCWMVKPGI-UHFFFAOYSA-N
2-bromo-3-bromomethyl pyridine, 2-bromo-3-bromomethylene pyridine, pubchem22284, 2-bromo-3-bromomethyl-pyridine, pyridine, 2-bromo-3-bromomethyl
10901078
2-bromo-3-(bromomethyl)pyridine
C1=CC(=C(N=C1)Br)CBr

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 38°C
Boiling Point 291°C
Flash Point >110°C (230°F)
Quantity 250 mg
Formula Weight 250.92
Percent Purity 96%
Chemical Name or Material 2-Bromo-3-(bromomethyl)pyridine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'