missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-3,5-difluorophenol, 98%

Product Code. 11366619
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11366619 1 g 1g
11376619 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11366619 Supplier Thermo Scientific Alfa Aesar Supplier No. H26203.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 325486-43-9 | C6H3BrF2O | 208.99 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 325486-43-9
Molecular Formula C6H3BrF2O
Molecular Weight (g/mol) 208.99
MDL Number MFCD01861127
InChI Key AGMQCXYWIMAOMT-UHFFFAOYSA-N
Synonym acmc-20a5l1, phenol,2-bromo-3,5-difluoro, phenol, 2-bromo-3,5-difluoro, 2-bromanyl-3,5-bis fluoranyl phenol
PubChem CID 22144860
IUPAC Name 2-bromo-3,5-difluorophenol
SMILES C1=C(C=C(C(=C1F)Br)O)F

Specifications

Refractive Index 1.529
Quantity 1 g
Formula Weight 208.99
Percent Purity 98%
Chemical Name or Material 2-Bromo-3,5-difluorophenol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.