missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Bromo-2-propenoic acid, TRC
Click to view available options
Quantity:
1 g
Unit Size:
1g
Specifications
Specifications
| Chemical Name or Material | 2-Bromo-2-propenoic acid |
| CAS | 10443-65-9 |
| Molecular Formula | C3H3BrO2 |
| Quantity | 1 g |
| InChI Formula | InChI=1S/C3H3BrO2/c1-2(4)3(5)6/h1H2,(H,5,6) |
| SMILES | OC(=O)C(=C)Br |
| IUPAC Name | 2-bromoprop-2-enoic acid |
| Molecular Weight (g/mol) | 150.96 |
| Formula Weight | 149.9316 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction