missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-2-nitropropane, 98%

Product Code. 11428317
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11428317 5 g 5g
11438317 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11428317 Supplier Thermo Scientific Alfa Aesar Supplier No. A18385.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 5447-97-2 | C3H6BrNO2 | 167.99 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5447-97-2
Molecular Formula C3H6BrNO2
Molecular Weight (g/mol) 167.99
MDL Number MFCD00007389
InChI Key OADSZWXMXIWZSQ-UHFFFAOYSA-N
Synonym propane, 2-bromo-2-nitro, 2-bromo-2-nitro-propane, snxhevlqiijip@, acmc-1aqp4, propane,2-bromo-2-nitro, 2-bromo-2-nitropropane, oadszwxmxiwzsq-uhfffaoysa
PubChem CID 79531
IUPAC Name 2-bromo-2-nitropropane
SMILES CC(C)([N+](=O)[O-])Br

Specifications

Density 1.584
Boiling Point 45°C to 46°C (16 mmHg)
Flash Point 61°C (141°F)
Refractive Index 1.464
Quantity 5 g
Beilstein 1746626
Formula Weight 168
Percent Purity 98%
Chemical Name or Material 2-Bromo-2-nitropropane

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.