missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-2-methylpropane, TRC

Product Code. 30337045
Change view
Click to view available options
Quantity:
2.5 g
Unit Size:
2.5g
Product Code. Quantity unitSize
30337045 2.5 g 2.5g
1 options
This item is not returnable. View return policy

Product Code. 30337045

Brand: Toronto Research Chemicals TRCB6789332.5G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-Bromo-2-methylpropane
CAS 507-19-7
Molecular Formula C4H9Br
Quantity 2.5 g
Synonym Propane, 2-bromo-2-methyl- (8CI, 9CI, ACI), 2-Bromo-2-methylpropane (ACI), 1,1-Dimethylethyl bromide, 2-Bromoisobutane, 2-Methyl-2-bromopropane, Bromotrimethylmethane, NSC 8418, tert-Butyl bromide, Trimethylbromomethane
InChI Formula InChI=1S/C4H9Br/c1-4(2,3)5/h1-3H3
SMILES CC(C)(C)Br
IUPAC Name 2-bromo-2-methylpropane
Molecular Weight (g/mol) 137.02
Formula Weight 135.9888
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.