missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-2-methylpropane, TRC

Product Code. 30331709
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30331709 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30331709 Supplier Toronto Research Chemicals Supplier No. TRCB678933500MG

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.
, 02 950 59 478

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-Bromo-2-methylpropane
CAS 507-19-7
Molecular Formula C4H9Br
Quantity 500 mg
Synonym Propane, 2-bromo-2-methyl- (8CI, 9CI, ACI), 2-Bromo-2-methylpropane (ACI), 1,1-Dimethylethyl bromide, 2-Bromoisobutane, 2-Methyl-2-bromopropane, Bromotrimethylmethane, NSC 8418, tert-Butyl bromide, Trimethylbromomethane
InChI Formula InChI=1S/C4H9Br/c1-4(2,3)5/h1-3H3
SMILES CC(C)(C)Br
IUPAC Name 2-bromo-2-methylpropane
Molecular Weight (g/mol) 137.02
Formula Weight 135.9888
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.