missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Bromo-2'-methylacetophenone, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
Description
2-Bromo-2'-methylacetophenone is an aromatic building block for proteomics research.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Bromo-2′-methylacetophenone is an aromatic building block for proteomics research.
Solubility
Slightly soluble in water.
Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
2-Bromo-2′-methylacetophenone is an aromatic building block for proteomics research.
Solubility
Slightly soluble in water.
Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
Specifications
Boiling Point | 81°C to 83°C |
Quantity | 1 g |
Beilstein | 508116 |
Solubility Information | Slightly soluble in water. |
Formula Weight | 213.07 |
Percent Purity | 98% |
Chemical Name or Material | 2-Bromo-2'-methylacetophenone |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'