missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-2'-methylacetophenone, 98%

Product Code. 15491939
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15491939 5 g 5g
15481939 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15491939 Supplier Thermo Scientific Alfa Aesar Supplier No. H64651.06
 Restricted Item
Estimated Shipment: 21-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 51012-65-8 | C9H9BrO | 213.074 g/mol

2-Bromo-2'-methylacetophenone is an aromatic building block for proteomics research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Bromo-2′-methylacetophenone is an aromatic building block for proteomics research.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 51012-65-8
Molecular Formula C9H9BrO
Molecular Weight (g/mol) 213.074
MDL Number MFCD04038955
InChI Key XMGAXELQRATLJP-UHFFFAOYSA-N
Synonym 2-bromo-1-o-tolyl ethanone, 2-bromo-1-2-methylphenyl ethan-1-one, 2'-methylphenacyl bromide, 2-bromo-1-o-tolylethanone, 2-bromo-1-2-methylphenyl ethanone, 2-methylphenacyl bromide, 2-bromo-1-o-tolyl-ethanone, 2-bromo-2'-methylacetophenone, ethanone, 2-bromo-1-methylphenyl, 2-bromoacetyl toluene
PubChem CID 12430676
IUPAC Name 2-bromo-1-(2-methylphenyl)ethanone
SMILES CC1=CC=CC=C1C(=O)CBr

Specifications

Boiling Point 81°C to 83°C
Quantity 5 g
Beilstein 508116
Solubility Information Slightly soluble in water.
Formula Weight 213.07
Percent Purity 98%
Chemical Name or Material 2-Bromo-2'-methylacetophenone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.