missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-2'-methylacetophenone, 98%, Thermo Scientific Chemicals

Catalog Number 15481939
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
51012-65-8
C9H9BrO
213.074
MFCD04038955
XMGAXELQRATLJP-UHFFFAOYSA-N
2-bromo-1-o-tolyl ethanone, 2-bromo-1-2-methylphenyl ethan-1-one, 2'-methylphenacyl bromide, 2-bromo-1-o-tolylethanone, 2-bromo-1-2-methylphenyl ethanone, 2-methylphenacyl bromide, 2-bromo-1-o-tolyl-ethanone, 2-bromo-2'-methylacetophenone, ethanone, 2-bromo-1-methylphenyl, 2-bromoacetyl toluene
12430676
2-bromo-1-(2-methylphenyl)ethanone
CC1=CC=CC=C1C(=O)CBr
This item is not returnable. View return policy
51012-65-8
C9H9BrO
213.074
MFCD04038955
XMGAXELQRATLJP-UHFFFAOYSA-N
2-bromo-1-o-tolyl ethanone, 2-bromo-1-2-methylphenyl ethan-1-one, 2'-methylphenacyl bromide, 2-bromo-1-o-tolylethanone, 2-bromo-1-2-methylphenyl ethanone, 2-methylphenacyl bromide, 2-bromo-1-o-tolyl-ethanone, 2-bromo-2'-methylacetophenone, ethanone, 2-bromo-1-methylphenyl, 2-bromoacetyl toluene
12430676
2-bromo-1-(2-methylphenyl)ethanone
CC1=CC=CC=C1C(=O)CBr

2-Bromo-2'-methylacetophenone is an aromatic building block for proteomics research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Bromo-2′-methylacetophenone is an aromatic building block for proteomics research.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY
Boiling Point 81°C to 83°C
Quantity 1 g
Beilstein 508116
Solubility Information Slightly soluble in water.
Formula Weight 213.07
Percent Purity 98%
Chemical Name or Material 2-Bromo-2'-methylacetophenone

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'