missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-2'-methoxyacetophenone, 98%

Product Code. 11392017
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11392017

Brand: Thermo Scientific Alfa Aesar L07696.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 31949-21-0 | C9H9BrO2 | 229.073 g/mol

2-Bromo-2'-methoxyacetophenone is used as a pharmaceutical intermediate .

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Bromo-2′-methoxyacetophenone is used as a pharmaceutical intermediate .

Solubility
Insoluble in water.

Notes
Keep container tightly closed. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 31949-21-0
Molecular Formula C9H9BrO2
Molecular Weight (g/mol) 229.073
MDL Number MFCD00000196
InChI Key GKNCPTLOPRDYMH-UHFFFAOYSA-N
Synonym 2-bromo-1-2-methoxyphenyl ethanone, 2-bromo-2'-methoxyacetophenone, 2-methoxyphenacyl bromide, o-methoxy phenacylbromide, bromomethyl 2-methoxyphenyl ketone, 2-bromo-1-2-methoxyphenyl ethan-1-one, 2-bromo-2'-methoxy acetophenone, ethanone, 2-bromo-1-methoxyphenyl, alpha-bromo-o-methoxyacetophenone, 2-bromoacetylanisole
PubChem CID 123440
IUPAC Name 2-bromo-1-(2-methoxyphenyl)ethanone
SMILES COC1=CC=CC=C1C(=O)CBr

Specifications

Melting Point 42°C to 46°C
Boiling Point 130°C (1 mmHg)
Flash Point >110°C (230°F)
Quantity 1 g
UN Number UN3261
Beilstein 1938788
Solubility Information Insoluble in water.
Formula Weight 229.08
Percent Purity 98%
Chemical Name or Material 2-Bromo-2'-methoxyacetophenone

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.