missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals 2'-Bromo-2'-deoxyuridine, 98%

Product Code. 15416539
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15416539 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15416539 Supplier Thermo Scientific Chemicals Supplier No. J64704.ME

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 72218-68-9 | C9H11BrN2O5 | 307.10 g/mol

2'-Bromo-2'-deoxyuridine is a thymidine analogue which is incorporated into DNA. 5-BrdU is routinely and extensively used to measure DNA synthesis and to label dividing cells. Consequently 5-BrdU is used to study cell signaling and other processes that induce cell proliferation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2′-Bromo-2′-deoxyuridine is a thymidine analogue which is incorporated into DNA. 5-BrdU is routinely and extensively used to measure DNA synthesis and to label dividing cells. Consequently 5-BrdU is used to study cell si
TRUSTED_SUSTAINABILITY

Specifications

CAS 72218-68-9
Color White
Molecular Formula C9H11BrN2O5
MDL Number MFCD06658226
Quantity 500 mg
Synonym 2'-bromo-2'-deoxyuridine, 2'-bromo-2'-deoxy-d-uridine, 2'-deoxy-2'-bromo-uridine, 1-2r,3r,4r,5r-3-bromo-4-hydroxy-5-hydroxymethyl tetrahydrofuran-2-yl pyrimidine-2,4 1h,3h-dione, 1-2r,3r,4r,5r-3-bromo-4-hydroxy-5-hydroxymethyl oxolan-2-yl-3h-pyrimidine-2,4-dione, 2'-bromodesoxyuridine, 2'-bromo-2'-deoxyuridin, 2-bromo-2-deoxyuridine, 1-2r,3r,4r,5r-3-bromo-4-hydroxy-5-hydroxymethyl tetrahydrofuran-2-yl pyrimidine-2,4-dione
Solubility Information Soluble in water.
InChI Key FEUDNSHXOOLCEY-XVFCMESISA-N
SMILES OC[C@H]1O[C@H]([C@H](Br)[C@@H]1O)N1C=CC(=O)NC1=O
IUPAC Name 1-[(2R,3R,4R,5R)-3-bromo-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-1,2,3,4-tetrahydropyrimidine-2,4-dione
Molecular Weight (g/mol) 307.10
PubChem CID 188257
Formula Weight 307.1
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 2'-Bromo-2'-deoxyuridine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.