missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-2'-chloroacetophenone, 95%

Product Code. 10336694 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10336694 1 g Glass bottle 1g
10346694 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10336694 Supplier Thermo Scientific Acros Supplier No. 427890010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 5000-66-8 | C8H6BrClO | 233.49 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5000-66-8
Molecular Formula C8H6BrClO
Molecular Weight (g/mol) 233.49
MDL Number MFCD00832993
InChI Key WZWWEVCLPKAQTA-UHFFFAOYSA-N
Synonym 2-bromo-2'-chloroacetophenone, 2-bromo-1-2-chlorophenyl ethanone, 2-bromo-1-2-chlorophenyl ethan-1-one, 2'-chlorophenacyl bromide, 2-chlorophenacyl bromide, 2'-chloro-2-bromoacetophenone, bromomethyl 2-chlorophenyl ketone, 2-bromo-1-2-chlorophenyl-1-ethanone, zlchem 501, 2-chlorophenacylbromide
PubChem CID 2735785
IUPAC Name 2-bromo-1-(2-chlorophenyl)ethanone
SMILES C1=CC=C(C(=C1)C(=O)CBr)Cl

Specifications

CAS Min % 94.0
CAS Max % 100.0
Color Colorless to Yellow
Density 1.6020g/mL
Boiling Point 162°C to 166°C (20.0 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Linear Formula BrCH2C(O)C6H4Cl
Quantity 1 g
Specific Gravity 1.602
Formula Weight 233.49
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material 2-Bromo-2′-chloroacetophenone
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2-Bromo-2'-chloroacetophenone
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.