missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-2',4'-difluoroacetophenone, 97%, Thermo Scientific™

Product Code. 15482918
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15482918 1 g 1g
15492918 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15482918 Supplier Thermo Scientific Alfa Aesar Supplier No. H56442.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 102429-07-2
Molecular Formula C8H5BrF2O
Molecular Weight (g/mol) 235.028
MDL Number MFCD00142822
InChI Key CSGDTHXBRAAOHV-UHFFFAOYSA-N
Synonym 2-bromo-2',4'-difluoroacetophenone, 2-bromo-1-2,4-difluorophenyl ethanone, 2,4-difluorophenacyl bromide, 2-bromo-1-2,4-difluorophenyl ethan-1-one, 2-bromo-1-2,4-difluoro-phenyl-ethanone, 2',4'-difluorophenacyl bromide, 2-bromo-2,4-difluoroacetophenone, ethanone, 2-bromo-1-2,4-difluorophenyl, alpha-bromo-2',4'-difluoroacetophenone, acmc-1c4aw
PubChem CID 2774185
IUPAC Name 2-bromo-1-(2,4-difluorophenyl)ethanone
SMILES C1=CC(=C(C=C1F)F)C(=O)CBr

Specifications

Melting Point 31°C to 35°C
Flash Point 51°C (124°F)
Quantity 1 g
UN Number UN2925
Sensitivity Air sensitive
Formula Weight 235.03
Percent Purity 97%
Chemical Name or Material 2-Bromo-2',4'-difluoroacetophenone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.