missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-2',4'-dichloroacetophenone, TRC

Product Code. 30347270
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30347270 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30347270 Supplier Toronto Research Chemicals Supplier No. TRCB6834055G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-Bromo-2',4'-dichloroacetophenone
CAS 2631-72-3
Molecular Formula C8H5BrCl2O
Quantity 5 g
Synonym 2,4-Dichlorophenacyl bromide, 2,4-Dichlorophenyl Bromomethyl Ketone, 2-Bromo-1-(2,4-dichlorophenyl)ethanone, 2',4'-Dichloro-2-bromoacetophenone, 2-Bromo-1-(2,4-dichlorophenyl)ethan-1-one, alpha-Bromo-2,4-dichloroacetophenone, ω-Bromo-2,4-dichloroacetophenone
InChI Formula InChI=1S/C8H5BrCl2O/c9-4-8(12)6-2-1-5(10)3-7(6)11/h1-3H,4H2
SMILES Clc1ccc(C(=O)CBr)c(Cl)c1
IUPAC Name 2-bromo-1-(2,4-dichlorophenyl)ethanone
Molecular Weight (g/mol) 267.93
Formula Weight 265.8901
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.