missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-2',4'-dichloroacetophenone, 97%, Thermo Scientific™

Product Code. p-2716094 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10726242 5 g Glass bottle 5g
10347701 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10726242 Supplier Thermo Scientific Acros Supplier No. 344990050

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2631-72-3
Molecular Formula C8H5BrCl2O
Molecular Weight (g/mol) 267.931
MDL Number MFCD00053005
InChI Key DASJDMQCPIDJIF-UHFFFAOYSA-N
Synonym 2-bromo-2',4'-dichloroacetophenone, 2-bromo-1-2,4-dichlorophenyl ethanone, 2,4-dichlorophenacyl bromide, 2-bromo-1-2,4-dichlorophenyl ethan-1-one, 2-bromo-1-2,4-dichloro-phenyl-ethanone, ethanone, 2-bromo-1-2,4-dichlorophenyl, 2'-bromo-2,4-dichloro acetophenone, omega-bromo-2,4-dichloro acetophenone, 2',4'-dichlorophenacyl bromide, 2-bromo-2,4-dichloroacetophenone
PubChem CID 75828
IUPAC Name 2-bromo-1-(2,4-dichlorophenyl)ethanone
SMILES C1=CC(=C(C=C1Cl)Cl)C(=O)CBr

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color White to Brown
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Quantity 5 g
Formula Weight 267.94
Percent Purity 97%
Physical Form Low Melting Solid
Chemical Name or Material 2-Bromo-2', 4'-dichloroacetophenone, 98%
Show More Show Less
compliance-icons
Product Identifier
  • 2-Bromo-2',4'-dichloroacetophenone
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.