missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-1H-benzimidazole, 97%, Maybridge

Product Code. 10202592
Change view
Click to view available options
Amber Glass Bottle; 1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10202592 Amber Glass Bottle; 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10202592 Supplier Thermo Scientific Maybridge Supplier No. CC09010DA

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 54624-57-6
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Molecular Formula C7H5BrN2
MDL Number MFCD00160009
Quantity 1 g
Synonym 2-bromobenzimidazole, 2-bromo-1h-benzo d imidazole, 2-bromo-1h-1,3-benzodiazole, 2-bromo-1h-benzoimidazole, 1h-benzimidazole, 2-bromo, bromobenzimidazole, bromo-benzimidazole, pubchem16113, acmc-1an7o, 1h-benzimidazole,2-bromo
InChI Key PHPYXVIHDRDPDI-UHFFFAOYSA-N
SMILES C1=CC=C2C(=C1)NC(=N2)Br
IUPAC Name 2-bromo-1H-benzimidazole
Molecular Weight (g/mol) 197.035
PubChem CID 2776281
Formula Weight 197.03
Percent Purity 97%
Chemical Name or Material 2-Bromo-1H-benzimidazole
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.