missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-1-indanol, 99%, Thermo Scientific™

Product Code. 15342022 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15342022 250 g 250g
15322022 10 g 10g
15332022 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15342022 Supplier Thermo Scientific Acros Supplier No. 107052500
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 5400-80-6
Molecular Formula C9H9BrO
Molecular Weight (g/mol) 213.07
MDL Number MFCD00003798
InChI Key RTESDSDXFLYAKZ-UHFFFAOYNA-N
Synonym 2-bromo-1-indanol, 2-bromoindan-1-ol, 1-hydroxy-2-bromoindan, 2-bromo-1-hydroxyindane, 2-bromo-1-hydroxyhydrindene, 1h-inden-1-ol, 2-bromo-2,3-dihydro, trans-2-bromo-1-indanol, bromoindanol, acmc-20ajgx, pubchem3783
PubChem CID 95444
IUPAC Name 2-bromo-2,3-dihydro-1H-inden-1-ol
SMILES OC1C(Br)CC2=CC=CC=C12

Specifications

Melting Point 130.0°C to 131.0°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Quantity 250 g
Formula Weight 213.07
Percent Purity 99%
Chemical Name or Material 2-Bromo-1-indanol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.